• USA Home
  • Search Results

  • purewaterdensitystandard1802773218511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pure Water Density Standard

2 Products
Synonym:   Water
Image of Pure Water Density Standard
CAS Number:
Linear Formula:
 H2O
Molecular Weight:
 18.02
 Beilstein/REAXYS Number:
 2050024
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Density
  Anal. Standard Form
  Shelf Life
  Manufacturer designation
Show More