• USA Home
  • Search Results

  • pyrazinamide123119896411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pyrazinamide

4 Products
Synonym:   Pyrazinamide, Pyrazinecarboxamide, Pyrazinoic acid amide
Image of Pyrazinamide
CAS Number:
Empirical Formula (Hill Notation):
 C5H5N3O
Molecular Weight:
 123.11
 Beilstein/REAXYS Number:
 112306
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
Show More