• USA Home
  • Search Results

  • pyrene2022512900011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pyrene

7 Products
Synonym:   Benzo[def]phenanthrene
Image of Pyrene
CAS Number:
Empirical Formula (Hill Notation):
 C16H10
Molecular Weight:
 202.25
 Beilstein/REAXYS Number:
 1307225
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Purity Grade
  Melting Point
  Fluorescence Test
  Anal. Standard Form
  Physical Form
  Special Grade
  Assay
Show More