• USA Home
  • Search Results

  • pyridine791011086111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pyridine

17 Products
Image of Pyridine
CAS Number:
Empirical Formula (Hill Notation):
 C5H5N
Molecular Weight:
 79.10
 Beilstein/REAXYS Number:
 103233
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  UV-Absorption Condition
  Purity Grade
  Ice Shipping
  Autoignition temp
  Agency/Method
  Vapor pressure
  Transition temperature
  Product Line
  Package quantity
  Melting Point
  Density
  Color
  Potency
  UV-Absorption Values
  Solubility
  Application(s)
  Special Grade
  Physical Form
  pH Value
Show More

Product #
Description
270970 Pyridine, anhydrous, 99.8%,
360570 Pyridine, ACS reagent, ≥99.0%,
270407 Pyridine, for HPLC, ≥99.9%,
1.09728 Pyridine, for analysis EMSURE® ACS,Reag. Ph Eur,
P57506 Pyridine, ReagentPlus®, ≥99%,
PX2020 Pyridine, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
W296600 Pyridine, ≥99%,
02486 Pyridine, analytical standard,
33553 Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC),
PHR1558 Pyridine, Pharmaceutical Secondary Standard; Certified Reference Material,
494410 Pyridine, biotech. grade, ≥99.9%,
320498 Pyridine, ReagentPlus®, ≥99%,
1.07462 Pyridine, EMPLURA®,
PX2014 Pyridine, OmniSolv®,
676772 Pyridine, ACS reagent, ≥99.0%,
L093000 Pyridine,
PX2012 Pyridine, anhydrous, septum-sealed bottle DriSolv,