• USA Home
  • Search Results

  • pyridiniumptoluenesulfonate251302405728111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pyridinium p-toluenesulfonate

2 Products
Synonym:   p-Toluenesulfonic acid pyridine salt, PPTS, Pyridine p-toluenesulfonate
Image of Pyridinium <I>p</I>-toluenesulfonate
CAS Number:
Empirical Formula (Hill Notation):
 C12H13NO3S
Molecular Weight:
 251.30
 Beilstein/REAXYS Number:
 3764305
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More