• USA Home
  • Search Results

  • pyrroloquinolinequinone330217290934311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pyrroloquinoline quinone

2 Products
Synonym:   4,5-Dihydro-4,5-dioxo-1H-pyrrolo[2,3-f]quinoline-2,7,9-tricarboxylic acid, Methoxatin, PQQ
Image of Pyrroloquinoline quinone
CAS Number:
Empirical Formula (Hill Notation):
 C14H6N2O8
Molecular Weight:
 330.21
 Beilstein/REAXYS Number:
 3596812
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Color
Show More