• USA Home
  • Search Results

  • pyruvatecarboxylasefrombovineliver12345901419111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pyruvate Carboxylase from bovine liver

1 Products
Synonym:   Pyruvate:CO2 ligase (ADP-forming)
CAS Number:
 Enzyme Commission (EC) Number:
 6.4.1.1  ( BRENDA | IUBMB )
 MDL number:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More