• USA Home
  • Search Results

  • pyruvic13c3acid910437878577411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pyruvic-13C3 acid

2 Products
Synonym:   α-Ketopropionic acid-13C3, 2-Oxopropionic acid-13C3
Image of Pyruvic-<SUP>13</SUP>C<SUB>3 </SUB>acid
CAS Number:
Linear Formula:
 13CH313CO13COOH
Molecular Weight:
 91.04
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Isotopic Purity
Show More