• USA Home
  • Search Results

  • pyruvicacid880612717311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Pyruvic acid

6 Products
Synonym:   α-Ketopropionic acid, 2-Oxopropionic acid
Image of Pyruvic acid
CAS Number:
Linear Formula:
 CH3COCOOH
Molecular Weight:
 88.06
 Beilstein/REAXYS Number:
 506211
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
 eCl@ss:
 39021320
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Document Links
  Allergen
  Organoleptic property
  Density
  Solubility
  Agency/Method
  Special Grade
Show More