• USA Home
  • Search Results

  • quercetin3022411739511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quercetin

2 Products
Synonym:   2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 3,3′,4′,5,6-Pentahydroxyflavone
Image of Quercetin
CAS Number:
Empirical Formula (Hill Notation):
 C15H10O7
Molecular Weight:
 302.24
 Beilstein/REAXYS Number:
 317313
 EC Number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More