• USA Home
  • Search Results

  • quercetin3gentiobioside62652743183611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quercetin 3-gentiobioside

1 Products
Synonym:   3,3′,4′,5,7-Pentahydroxyflavone 3-gentiobioside, Quercetin 3-O-β-gentiobioside
Image of Quercetin 3-gentiobioside
CAS Number:
Empirical Formula (Hill Notation):
 C27H30O17
Molecular Weight:
 626.52
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More