• USA Home
  • Search Results

  • quercetin3oalarabinopyranoside434352225513611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quercetin 3-O-α-L-arabinopyranoside

1 Products
Synonym:   3′,4′,5,7-Tetrahydroxyflavone 3-O-α-L-arabinoside, 3-(α-L-Arabinopyranosyloxy)-2-(3,4-dihydroxyphenyl)-5,7-dihydroxy-4H-1-benzopyran-4-one, Feniculin, Foeniculin, Guaijaverin, Quercetin 3-O-α-L-arabinoside
Image of Quercetin 3-<I>O</I>-α-<SC>L</SC>-arabinopyranoside
CAS Number:
Empirical Formula (Hill Notation):
 C20H18O11
Molecular Weight:
 434.35
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More