• USA Home
  • Search Results

  • quercetin3orutinoside7oglucoside772663031161611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quercetin 3-O-rutinoside-7-O-glucoside

2 Products
Synonym:   3,3′,4′,5,7-Pentahydroxyflavone 3-rutinoside 7-glucoside, 3-[[6-O-(6-Deoxy-α-L-mannopyranosyl)-β-D-glucopyranosyl]oxy]-2-(3,4-dihydroxyphenyl)-7-(β-D-glucopyranosyloxy)-5-hydroxy-4H-1-benzopyran-4-one, Quercetin 3-rhamnoglucoside 7-glucoside
Image of Quercetin 3-<I>O</I>-rutinoside-7-<I>O</I>-glucoside
CAS Number:
Empirical Formula (Hill Notation):
 C33H40O21
Molecular Weight:
 772.66
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
Show More