• USA Home
  • Search Results

  • quinacrinedihydrochloride472886905611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quinacrine dihydrochloride

2 Products
Synonym:   6-Chloro-9-(4-diethylamino-1-methylbutylamino)-2-methoxyacridine dihydrochloride, Atebrin dihydrochloride, Mepacrine dihydrochloride
Image of Quinacrine dihydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C23H30ClN3O · 2HCl
Molecular Weight:
 472.88
 Beilstein/REAXYS Number:
 4834013
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Solubility
Show More