• USA Home
  • Search Results

  • quinazoline1301525382711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quinazoline

1 Products
Synonym:   1,3-Benzodiazine, Benzopyrimidine
Image of Quinazoline
CAS Number:
Empirical Formula (Hill Notation):
 C8H6N2
Molecular Weight:
 130.15
 Beilstein/REAXYS Number:
 109370
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
Show More

Product #
Description
123323 Quinazoline, 99%,