• USA Home
  • Search Results

  • quinicacid192177795211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(−) Quinic acid

3 Products
Synonym:   D-(−)-Quinic acid
Image of (−) Quinic acid
CAS Number:
Empirical Formula (Hill Notation):
 C7H12O6
Molecular Weight:
 192.17
 Beilstein/REAXYS Number:
 2212412
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
Show More