• USA Home
  • Search Results

  • quinidine324425654211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quinidine

2 Products
Image of Quinidine
CAS Number:
Empirical Formula (Hill Notation):
 C20H24N2O2
Molecular Weight:
 324.42
 Beilstein/REAXYS Number:
 91866
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More