• USA Home
  • Search Results

  • quinidinesulfate78294659163511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quinidine sulfate

2 Products
Synonym:   Quinidine sulfate salt dihydrate
Image of Quinidine sulfate
CAS Number:
Empirical Formula (Hill Notation):
 C40H48N4O4 · H2O4S · 2H2O
Molecular Weight:
 782.94
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Manufacturer designation
  Special Grade
Show More