• USA Home
  • Search Results

  • quinidinesulfatesaltdihydrate78294659163511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quinidine sulfate salt dihydrate

1 Products
Image of Quinidine sulfate salt dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C40H48N4O4 · H2O4S · 2H2O
Molecular Weight:
 782.94
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More