• USA Home
  • Search Results

  • quininehemisulfatesaltmonohydrate3914720767144111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quinine hemisulfate salt monohydrate

3 Products
Synonym:   Quinine sulfate (2:1) (salt) dihydrate
Image of Quinine hemisulfate salt monohydrate
CAS Number:
Empirical Formula (Hill Notation):
 C20H24N2O2 · 0.5H2O4S · H2O
Molecular Weight:
 391.47
 Beilstein/REAXYS Number:
 6113937
 MDL number:
 NACRES:
 NA.32
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Solubility
  Assay
  Mode of Action
Show More