• USA Home
  • Search Results

  • quininehydrochloridedihydrate39691611947711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quinine hydrochloride dihydrate

5 Products
Synonym:   (8α, 9R)-6′-Methoxycinchonan-9-ol monohydrochloride dihydrate
Image of Quinine hydrochloride dihydrate
CAS Number:
Empirical Formula (Hill Notation):
 C20H24N2O2 · HCl · 2H2O
Molecular Weight:
 396.91
 Beilstein Registry Number:
 6112655
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
  Fluorescence Test
  Solubility
Show More