• USA Home
  • Search Results

  • quinoline129169122511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quinoline

4 Products
Synonym:   1-Benzazine, 2,3-Benzopyridine
Image of Quinoline
CAS Number:
Empirical Formula (Hill Notation):
 C9H7N
Molecular Weight:
 129.16
 Beilstein/REAXYS Number:
 107477
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Application(s)
  Potency
  Restriction
  Special Grade
Show More