• USA Home
  • Search Results

  • quinolineyellow12345800492011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Quinoline Yellow

2 Products
Synonym:   Acid Yellow 3
Image of Quinoline Yellow
CAS Number:
 Colour Index Number:
 47005
 NACRES:
 NA.47
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Absorption
  Physical Form
  Solubility
Show More