• USA Home
  • Search Results

  • r12propanediol7609425414211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(−)-1,2-Propanediol

1 Products
Synonym:   (R)-(−)-Propylene glycerol, (R)-(−)-Propylene glycol
Image of (<I>R</I>)-(−)-1,2-Propanediol
CAS Number:
Linear Formula:
 CH3CH(OH)CH2OH
Molecular Weight:
 76.09
 Beilstein/REAXYS Number:
 1718872
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More