• USA Home
  • Search Results

  • r19anthryl222trifluoroethanol276255353134311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(−)-1-(9-Anthryl)-2,2,2-trifluoroethanol

1 Products
Synonym:   (R)-(−)-α-(Trifluoromethyl)anthracene-9-methanol, (R)-(−)-2,2,2-Trifluoro-1-(9-anthryl)ethanol
Image of (<I>R</I>)-(−)-1-(9-Anthryl)-2,2,2-trifluoroethanol
CAS Number:
Empirical Formula (Hill Notation):
 C16H11F3O
Molecular Weight:
 276.25
 Beilstein/REAXYS Number:
 4695163
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More