• USA Home
  • Search Results

  • r2oxothiazolidine4carboxylicacid147151977163211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(−)-2-Oxothiazolidine-4-carboxylic acid

1 Products
Synonym:   L-(−)-2-Oxothiazolidine-4-carboxylic acid
Image of (<I>R</I>)-(−)-2-Oxothiazolidine-4-carboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C4H5NO3S
Molecular Weight:
 147.15
 Beilstein/REAXYS Number:
 4179169
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More