• USA Home
  • Search Results

  • r3piperidinecarboxylicacid129162513700211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(–)-3-Piperidinecarboxylic acid

1 Products
Synonym:   (R)-(–)-Nipecotic acid
Image of (<I>R</I>)<WBR>-<WBR>(–)<WBR>-<WBR>3-<WBR>Piperidinecarboxylic acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H11NO2
Molecular Weight:
 129.16
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More