• USA Home
  • Search Results

  • rD12naphthylethanol172225219385811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(+)-1-(2-Naphthyl)ethanol

1 Products
Synonym:   (R)-(+)-α-Methyl-2-naphthalenemethanol
Image of (<I>R</I>)<WBR>-<WBR>(+)<WBR>-<WBR>1-<WBR>(2-<WBR>Naphthyl)<WBR>ethanol
CAS Number:
Linear Formula:
 C10H7CH(CH3)OH
Molecular Weight:
 172.22
 Beilstein/REAXYS Number:
 3195952
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More