• USA Home
  • Search Results

  • rDalipoicacid20633120022211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(+)-α-Lipoic acid

2 Products
Synonym:   (R)-6,8-Dithiooctanoic acid, (R)-6,8-Thioctic acid, (R)-(+)-1,2-Dithiolane-3-pentanoic acid
Image of (<I>R</I>)-(+)-α-Lipoic acid
CAS Number:
Empirical Formula (Hill Notation):
 C8H14O2S2
Molecular Weight:
 206.33
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More