• USA Home
  • Search Results

  • rDlimonene13623598927511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(+)-Limonene

7 Products
Synonym:   (+)-p-Mentha-1,8-diene, (+)-Carvene, (R)-4-Isopropenyl-1-methyl-1-cyclohexene
Image of (<I>R</I>)-(+)-Limonene
CAS Number:
Empirical Formula (Hill Notation):
 C10H16
Molecular Weight:
 136.23
 Beilstein/REAXYS Number:
 2204754
 EC Number:
 MDL number:
 NACRES:
 NA.79
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Shelf Life
  Vapor pressure
  Titertest
  Purity Grade
  Potency
  Density
  Special Grade
Show More