• USA Home
  • Search Results

  • rDpropyleneoxide58081544847211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(+)-Propylene oxide

1 Products
Synonym:   (R)-(+)-Methyloxirane, (R)-(+)-1,2-Epoxypropane
Image of (<I>R</I>)-(+)-Propylene oxide
CAS Number:
Empirical Formula (Hill Notation):
 C3H6O
Molecular Weight:
 58.08
 Beilstein/REAXYS Number:
 79764
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More