• USA Home
  • Search Results

  • rDpulegone152238982711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(+)-Pulegone

4 Products
Synonym:   (+)-Pulegone, (R)-p-Menth-4(8)-en-3-one, (R)-2-Isopropylidene-5-methylcyclohexanone, p-Menth-4(8)-en-3-one
Image of (<I>R</I>)-(+)-Pulegone
CAS Number:
Empirical Formula (Hill Notation):
 C10H16O
Molecular Weight:
 152.23
 Beilstein/REAXYS Number:
 2040703
 Council of Europe no.:
 2050c
 EC Number:
 FEMA Number:
 2963
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Potency
Show More