• USA Home
  • Search Results

  • rDstyreneoxide120152078053411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(+)-Styrene oxide

2 Products
Synonym:   (R)-(+)-Phenyloxirane, (R)-Phenylethylene oxide
Image of (<I>R</I>)-(+)-Styrene oxide
CAS Number:
Empirical Formula (Hill Notation):
 C8H8O
Molecular Weight:
 120.15
 Beilstein/REAXYS Number:
 1984
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Optical purity
Show More