• USA Home
  • Search Results

  • raneynickel12345744002011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Raney®-Nickel

4 Products
Synonym:   Nickel sponge, Raney Ni
Image of Raney<SUP>®</SUP>-Nickel
CAS Number:
Linear Formula:
 AlNi
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Reaction Suitability
  Physical Form
  Manufacturer designation
Show More