• USA Home
  • Search Results

  • ranitidinehydrochloride350866635759311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ranitidine hydrochloride

5 Products
Image of Ranitidine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C13H22N4O3S · HCl
Molecular Weight:
 350.86
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Solubility
  Anal. Standard Form
  Special Grade
Show More