• USA Home
  • Search Results

  • rapamycinfromstreptomyceshygroscopicus914175312388911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rapamycin from Streptomyces hygroscopicus

1 Products
Synonym:   23,27-Epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine, AY 22989, Sirolimus
Image of Rapamycin from <I>Streptomyces hygroscopicus</I>
CAS Number:
Empirical Formula (Hill Notation):
 C51H79NO13
Molecular Weight:
 914.17
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Mode of Action
  Activity Spectrum
  Solubility
Show More