• USA Home
  • Search Results

  • reineckesalt336431357316511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Reinecke salt

2 Products
Synonym:   Ammonium reineckate, Ammonium tetrarhodanatodiamminechromate(III), Ammonium tetrathiocyanatodiamminechromate(III), Ammonium tetrathiocyanodiammonochromate
Image of Reinecke salt
CAS Number:
Linear Formula:
 NH4[Cr(NH3)2(SCN)4]
Molecular Weight:
 336.43
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
Show More