• USA Home
  • Search Results

  • resazurinsodiumsalt251176275813811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Resazurin sodium salt

2 Products
Synonym:   7-Hydroxy-3H-phenoxazin-3-one-10-oxide sodium salt
Image of Resazurin sodium salt
CAS Number:
Empirical Formula (Hill Notation):
 C12H6NNaO4
Molecular Weight:
 251.17
 Beilstein/REAXYS Number:
 3794971
 EC Number:
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Chemical Composition
  Solubility
  Absorption
Show More