• USA Home
  • Search Results

  • residualsolventacetonitrile41057505811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Residual Solvent - Acetonitrile

1 Products
Synonym:   ACN, Acetonitrile, Cyanomethane, Ethyl nitrile, Methyl cyanide
Image of Residual Solvent - Acetonitrile
CAS Number:
Linear Formula:
 CH3CN
Molecular Weight:
 41.05
 Beilstein/REAXYS Number:
 741857
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More