• USA Home
  • Search Results

  • residualsolventclass2toluene1234510888311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Residual Solvent Class 2 - Toluene

1 Products
Synonym:   Toluene in dimethyl sulfoxide, Toluene solution
CAS Number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More