• USA Home
  • Search Results

  • residualsolventtoluene921410888311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Residual Solvent - Toluene

1 Products
Synonym:   Toluene
Image of Residual Solvent - Toluene
CAS Number:
Linear Formula:
 C6H5CH3
Molecular Weight:
 92.14
 Beilstein/REAXYS Number:
 635760
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More