• USA Home
  • Search Results

  • resorcinol1101110846311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Resorcinol

13 Products
Synonym:   1,3-Benzenediol
Image of Resorcinol
CAS Number:
Linear Formula:
 C6H4-1,3-(OH)2
Molecular Weight:
 110.11
 Beilstein/REAXYS Number:
 906905
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Suitability
  Agency/Method
  Anal. Standard Form
  pH Value
  Vapor pressure
  Transition temperature
  Manufacturer designation
  Melting Point
  Explosion limit
  Density
  Bulk Density
  Autoignition temp
  Application(s)
  Product Line
  Potency
  Solubility
  Physical Form
  Special Grade
Show More