• USA Home
  • Search Results

  • resorufinpentylether283328768703411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Resorufin pentyl ether

1 Products
Synonym:   7-Pentyloxy-3-phenoxazone, 7-Pentyloxy-3H-phenoxazin-3-one, O7-Pentylresorufin, Pentoxyresorufin
Image of Resorufin pentyl ether
CAS Number:
Empirical Formula (Hill Notation):
 C17H17NO3
Molecular Weight:
 283.32
 Beilstein/REAXYS Number:
 8394939
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
P0928 Resorufin pentyl ether,