• USA Home
  • Search Results

  • resveratrol2282450136011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Resveratrol

4 Products
Synonym:   3,4′,5-Trihydroxy-trans-stilbene, 5-[(1E)-2-(4-Hydroxyphenyl)ethenyl]-1,3-benzenediol
Image of Resveratrol
CAS Number:
Empirical Formula (Hill Notation):
 C14H12O3
Molecular Weight:
 228.24
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Shelf Life
  Solubility
  Assay
  Anal. Standard Form
  Special Grade
  Application(s)
Show More