• USA Home
  • Search Results

  • retinoicacid3004430279411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Retinoic acid

1 Products
Synonym:   ATRA, Tretinoin, Vitamin A acid, all-trans-Retinoic acid
Image of Retinoic acid
CAS Number:
Empirical Formula (Hill Notation):
 C20H28O2
Molecular Weight:
 300.44
 Beilstein/REAXYS Number:
 2057223
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More