• USA Home
  • Search Results

  • retinolpalmitate524867981211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Retinol palmitate

1 Products
Synonym:   Vitamin A palmitate, all trans-Retinol palmitate
CAS Number:
Empirical Formula (Hill Notation):
 C36H60O2
Molecular Weight:
 524.86
 EC Number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More