• USA Home
  • Search Results

  • rhein2842247843311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rhein

4 Products
Synonym:   4,5-Dihydroxyanthraquinone-2-carboxylic acid, 9,10-Dihydro-4,5-dihydroxy-9,10-dioxo-2-anthracenecarboxylic acid, Cassic acid, Chrysazin 3-carboxylic acid, Monorhein, Rheic acid, Rhubarb yellow
Image of Rhein
CAS Number:
Empirical Formula (Hill Notation):
 C15H8O6
Molecular Weight:
 284.22
 Beilstein/REAXYS Number:
 2222155
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Application(s)
  Assay
Show More