• USA Home
  • Search Results

  • rhenium18621744015511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rhenium

93 Products
Image of Rhenium
CAS Number:
Empirical Formula (Hill Notation):
 Re
Molecular Weight:
 186.21
 EC Number:
 MDL number:
 NACRES:
 NA.23
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Composite Dimension
  Weight
  Miscellaneous Specifications
  Particle Dimension
  Assay
  Physical Form
  Manufacturer designation
Show More

Product #
Description
204188 Rhenium, powder, 99.995% trace metals basis,
267317 Rhenium, foil, thickness 0.25 mm, 99.98% trace metals basis,
267279 Rhenium, powder, −100 mesh, ≥99.9% trace metals basis,
GF02111937 Rhenium, wire reel, 0.05m, diameter 1.0mm, annealed, 99.97%,
GF62345831 Rhenium, foil, thickness 0.025 mm, coil width 0.76 mm, length 0.5 m,
GF66233538 Rhenium, foil, thickness 0.25 mm, size 25 × 25 mm, purity 99.99%,
GF68288316 Rhenium, foil, thickness 0.05 mm, length 1 m, coil width 0.76 mm,
GF69380376 Rhenium, foil, thickness 0.05 mm, length 0.2 m, coil width 0.76 mm,
GF69388666 Rhenium, rod, length 100 mm, 1.5 mm diameter, purity 99.99%,
GF72546307 Rhenium, foil, thickness 0.05 mm, length 0.5 m, coil width 0.76 mm,
GF74450853 Rhenium, foil, thickness 0.03 mm, length 0.1 m, coil width 0.76 mm,
GF74812349 Rhenium, foil, thickness 0.04 mm, length 0.2 m, coil width 0.8 mm,
GF77190352 Rhenium, foil, thickness 0.035 mm, length 0.2 m, coil width 0.76 mm,
GF81143636 Rhenium, foil, thickness 0.04 mm, length 2 m, coil width 0.8 mm,
GF83521975 Rhenium, powder, 45 max. part. size (micron), weight 10 g, purity 99.99%,
GF88309851 Rhenium, foil, thickness 0.15 mm, size 25 × 25 mm, purity 99.99%,
GF88891509 Rhenium, foil, thickness 0.04 mm, length 2 m, coil width 0.7 mm,
GF92641608 Rhenium, foil, thickness 0.04 mm, length 0.1 m, coil width 0.8 mm,
GF92850543 Rhenium, foil, thickness 0.04 mm, length 1 m, coil width 0.8 mm,
GF93228880 Rhenium, foil, thickness 0.025 mm, length 1 m, coil width 0.76 mm,
GF94583199 Rhenium, foil, thickness 0.025 mm, length 0.1 m, coil width 0.76 mm,
GF94586440 Rhenium, foil, thickness 0.03 mm, length 0.5 m, coil width 0.76 mm,
GF95834728 Rhenium, foil, thickness 1.0 mm, diameter 12 mm, purity 99.99%,
GF99284451 Rhenium, foil, thickness 0.04 mm, length 0.1 m, coil width 0.7 mm,
GF01395604 Rhenium, foil, thickness 0.025 mm, length 5 m, coil width 0.76 mm,
GF02955045 Rhenium, foil, thickness 0.05 mm, length 0.1 m, coil width 0.76 mm,
GF06356695 Rhenium, foil, thickness 0.03 mm, length 0.2 m, coil width 0.76 mm,
GF10557288 Rhenium, foil, thickness 0.04 mm, length 0.2 m, coil width 0.7 mm,
GF12991653 Rhenium, foil, thickness 0.025 mm, size 25 × 25 mm, tolerance 0.25,
GF13149176 Rhenium, rod, 1.5 mm diameter, length 25 mm, purity 99.99%,
GF13196033 Rhenium, foil, thickness 0.03 mm, length 2 m, coil width 0.76 mm,
GF19660080 Rhenium, foil, thickness 0.04 mm, length 0.5 m, coil width 0.7 mm,
GF21333642 Rhenium, foil, thickness 0.125 mm, size 25 × 25 mm, purity 99.99%,
GF22898762 Rhenium, foil, thickness 0.025 mm, length 0.2 m, coil width 0.76 mm,
GF23305564 Rhenium, foil, thickness 0.03 mm, length 1 m, coil width 0.76 mm,
GF29804980 Rhenium, foil, thickness 0.05 mm, size 25 × 25 mm, purity 99.99%,
GF30097818 Rhenium, foil, thickness 1.0 mm, size 25 × 25 mm, purity 99.99%,
GF36039764 Rhenium, foil, thickness 0.05 mm, length 2 m, coil width 0.76 mm,
GF40205416 Rhenium, foil, thickness 0.5 mm, size 25 × 25 mm, purity 99.99%,
GF40322573 Rhenium, foil, thickness 0.20 mm, size 25 × 25 mm, purity 99.99%,
GF42268170 Rhenium, powder, max. particle size 45 micron, weight 5 g, purity 99.99%,
GF48794271 Rhenium, foil, thickness 0.035 mm, length 0.5 m, coil width 0.76 mm,
GF50583102 Rhenium, rod, 1.5 mm diameter, length 50 mm, purity 99.99%,
GF50732704 Rhenium, foil, thickness 0.04 mm, length 0.5 m, coil width 0.8 mm,
GF53611163 Rhenium, foil, thickness 0.025 mm, length 2 m, coil width 0.76 mm,
GF54217526 Rhenium, foil, thickness 0.035 mm, length 0.1 m, coil width 0.76 mm,
GF56774256 Rhenium, foil, thickness 0.0125 mm, size 25 × 25 mm, purity 99.99%,
GF57511133 Rhenium, foil, thickness 0.04 mm, length 1 m, coil width 0.7 mm,
GF02978309 Rhenium, wire reel, 0.025m, diameter 1.0mm, annealed, 99.97%,
GF94222458 Rhenium, wire reel, 0.05m, diameter 0.5mm, annealed, 99.97%,
GF04116158 Rhenium, wire reel, 0.1m, diameter 0.076mm, annealed, 99.97%,
GF21278640 Rhenium, wire reel, 0.1m, diameter 0.10mm, annealed, 99.97%,
GF92772737 Rhenium, wire reel, 0.1m, diameter 0.125mm, annealed, 99.97%,
GF00817466 Rhenium, wire reel, 0.1m, diameter 0.15mm, annealed, 99.97%,
GF60560513 Rhenium, wire reel, 0.1m, diameter 0.178mm, annealed, 99.97%,
GF57863931 Rhenium, wire reel, 0.1m, diameter 0.25mm, annealed, 99.97%,
GF80386869 Rhenium, wire reel, 0.1m, diameter 0.2mm, annealed, 99.97%,
GF90374441 Rhenium, wire reel, 0.1m, diameter 0.3mm, annealed, 99.97%,
GF77806367 Rhenium, wire reel, 0.1m, diameter 0.5mm, annealed, 99.97%,
GF63991929 Rhenium, wire reel, 0.1m, diameter 1.0mm, annealed, 99.97%,
GF14519156 Rhenium, wire reel, 0.2m, diameter 0.076mm, annealed, 99.97%,
GF59830139 Rhenium, wire reel, 0.2m, diameter 0.10mm, annealed, 99.97%,
GF96591875 Rhenium, wire reel, 0.2m, diameter 0.125mm, annealed, 99.97%,
GF02183217 Rhenium, wire reel, 0.2m, diameter 0.15mm, annealed, 99.97%,
GF70064658 Rhenium, wire reel, 0.2m, diameter 0.178mm, annealed, 99.97%,
GF32991486 Rhenium, wire reel, 0.2m, diameter 0.25mm, annealed, 99.97%,
GF48358435 Rhenium, wire reel, 0.2m, diameter 0.2mm, annealed, 99.97%,
GF96805884 Rhenium, wire reel, 0.2m, diameter 0.3mm, annealed, 99.97%,
GF25796010 Rhenium, wire reel, 0.2m, diameter 0.5mm, annealed, 99.97%,
GF98990050 Rhenium, wire reel, 0.2m, diameter 1.0mm, annealed, 99.97%,
GF14894163 Rhenium, wire reel, 0.5m, diameter 0.076mm, annealed, 99.97%,
GF78766745 Rhenium, wire reel, 0.5m, diameter 0.10mm, annealed, 99.97%,
GF73984797 Rhenium, wire reel, 0.5m, diameter 0.125mm, annealed, 99.97%,
GF72901324 Rhenium, wire reel, 0.5m, diameter 0.15mm, annealed, 99.97%,
GF58081945 Rhenium, wire reel, 0.5m, diameter 0.178mm, annealed, 99.97%,
GF23936104 Rhenium, wire reel, 0.5m, diameter 0.25mm, annealed, 99.97%,
GF44804325 Rhenium, wire reel, 0.5m, diameter 0.2mm, annealed, 99.97%,
GF00735353 Rhenium, wire reel, 0.5m, diameter 0.3mm, annealed, 99.97%,
GF24502115 Rhenium, wire reel, 0.5m, diameter 0.5mm, annealed, 99.97%,
GF61770225 Rhenium, wire reel, 1m, diameter 0.076mm, annealed, 99.97%,
GF45842733 Rhenium, wire reel, 1m, diameter 0.10mm, annealed, 99.97%,
GF48099112 Rhenium, wire reel, 1m, diameter 0.125mm, annealed, 99.97%,
GF12665460 Rhenium, wire reel, 1m, diameter 0.15mm, annealed, 99.97%,
GF60242723 Rhenium, wire reel, 1m, diameter 0.178mm, annealed, 99.97%,
GF92012745 Rhenium, wire reel, 1m, diameter 0.25mm, annealed, 99.97%,
GF00541167 Rhenium, wire reel, 1m, diameter 0.2mm, annealed, 99.97%,
GF91919290 Rhenium, wire reel, 1m, diameter 0.3mm, annealed, 99.97%,
GF12389834 Rhenium, wire reel, 2m, diameter 0.125mm, annealed, 99.97%,
GF83182620 Rhenium, wire reel, 2m, diameter 0.15mm, annealed, 99.97%,
GF30341146 Rhenium, wire reel, 2m, diameter 0.178mm, annealed, 99.97%,
GF01592513 Rhenium, wire reel, 2m, diameter 0.25mm, annealed, 99.97%,
GF67645301 Rhenium, wire reel, 2m, diameter 0.2mm, annealed, 99.97%,
459984 Rhenium, foil, thickness 1.0 mm, 99.98% trace metals basis,