• USA Home
  • Search Results

  • rhexahydromandelicacid158195358593611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(R)-(−)-Hexahydromandelic acid

1 Products
Synonym:   (R)-(−)-Cyclohexylhydroxyacetic acid
Image of (<I>R</I>)-(−)-Hexahydromandelic acid
CAS Number:
Linear Formula:
 C6H11CH(OH)CO2H
Molecular Weight:
 158.19
 Beilstein/REAXYS Number:
 3196809
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More