• USA Home
  • Search Results

  • rhodamine123380826266970911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Rhodamine 123

2 Products
Synonym:   2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid methyl ester
Image of Rhodamine 123
CAS Number:
Linear Formula:
 C21H17ClN2O3
Molecular Weight:
 380.82
 Beilstein/REAXYS Number:
 6030951
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Application(s)
Show More